1-Methyl-5-(piperidin-4-yl)pyridin-2(1H)-one hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321256-100MG
APOSOR321256-100MGEA 333 EUR
APOSOR321256-100MG
1-Methyl-5-(piperidin-4-yl)pyridin-2(1H)-one hydrochloride 97%
1-Methyl-5-(piperidin-4-yl)pyridin-2(1H)-one hydrochloride
Formel: C₁₁H₁₆N₂O·ClH
Molecular Weight: 228,718 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30099257
CAS-Nummer: 1137950-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR