N-Methyl-4-(piperidin-4-yl)benzamide hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321257-500MG
APOSOR321257-500MGEA 217 EUR
APOSOR321257-500MG
N-Methyl-4-(piperidin-4-yl)benzamide hydrochloride 97%
N-Methyl-4-(piperidin-4-yl)benzamide hydrochloride
Formel: C₁₃H₁₈N₂O·ClH
Molecular Weight: 254,756 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099258
CAS-Nummer: 2064217-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR