(6-(Piperidin-4-yl)pyridin-2-yl)methanol dihydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR321258-100MG
APOSOR321258-100MGEA 168 EUR
APOSOR321258-100MG
(6-(Piperidin-4-yl)pyridin-2-yl)methanol dihydrochloride 95%
(6-(Piperidin-4-yl)pyridin-2-yl)methanol dihydrochloride
Formel: C₁₁H₁₆N₂O·2ClH
Molecular Weight: 265,179 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099259
CAS-Nummer: 2244088-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR