tert-Butyl 4-(3-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR321259-100MG
APOSOR321259-100MGEA
132
EUR
APOSOR321259-100MG
tert-Butyl 4-(3-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate 95%
tert-Butyl 4-(3-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate
Formel:
C₁₈H₂₇NO₃ Molecular Weight: 305,412 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30099261 CAS-Nummer: 1853217-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...