7-Hydroxy-6-propyl-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one 95%

Lieferant: Apollo Scientific

Synonyme: 4-Hydroxy-3-propyl-1-azatricyclo[7.3.1.05,13]trideca-3,5(13),6,8-tetraen-2-one

OR32126-1G OR32126-5G
APOSOR32126-1GEA 180 EUR
APOSOR32126-1G APOSOR32126-5G
7-Hydroxy-6-propyl-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one 95%
7-Hydroxy-6-propyl-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one
Formel: C₁₅H₁₇NO₂
Molecular Weight: 243.31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03791142
CAS-Nummer: 57625-51-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR