tert-Butyl 4-(2-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR321260-100MG
APOSOR321260-100MGEA
91.5
EUR
APOSOR321260-100MG
tert-Butyl 4-(2-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate 97%
tert-Butyl 4-(2-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate
Formel:
C₁₈H₂₇NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD30099263 CAS-Nummer: 170837-78-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...