tert-Butyl 4-(4-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

OR321261-100MG
APOSOR321261-100MGEA 168 EUR
APOSOR321261-100MG
tert-Butyl 4-(4-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate 95%
tert-Butyl 4-(4-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate
Formel: C₁₈H₂₇NO₃
Molecular Weight: 305,412 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099265
CAS-Nummer: 1853217-74-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR