N-Methyl-4-(piperidin-3-yl)benzamide hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR321262-100MG
APOSOR321262-100MGEA 333 EUR
APOSOR321262-100MG
N-Methyl-4-(piperidin-3-yl)benzamide hydrochloride 95%
N-Methyl-4-(piperidin-3-yl)benzamide hydrochloride
Formel: C₁₃H₁₈N₂O·ClH
Molecular Weight: 254,756 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099266
CAS-Nummer: 1931126-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR