4-(2-Chloro-5-methoxyphenyl)piperidine hydrochloride 90%
Lieferant: Apollo Scientific
|
Achtung
|
OR321264-100MG
APOSOR321264-100MGEA
433
EUR
APOSOR321264-100MG
4-(2-Chloro-5-methoxyphenyl)piperidine hydrochloride 90%
4-(2-Chloro-5-methoxyphenyl)piperidine hydrochloride
Formel:
C₁₂H₁₆ClNO·ClH Molecular Weight: 262,175 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD30099267 CAS-Nummer: 1044773-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...