2,6-Dimethyl-3-(piperidin-3-yl)pyridine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321265-100MG
APOSOR321265-100MGEA 333 EUR
APOSOR321265-100MG
2,6-Dimethyl-3-(piperidin-3-yl)pyridine dihydrochloride 97%
2,6-Dimethyl-3-(piperidin-3-yl)pyridine dihydrochloride
Formel: C₁₂H₁₈N₂·2ClH
Molecular Weight: 263.207 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099269
CAS-Nummer: 2244084-77-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR