N-(4-(Piperidin-3-yl)phenyl)acetamide hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321266-100MG
APOSOR321266-100MGEA 168 EUR
APOSOR321266-100MG
N-(4-(Piperidin-3-yl)phenyl)acetamide hydrochloride 97%
N-(4-(Piperidin-3-yl)phenyl)acetamide hydrochloride
Formel: C₁₃H₁₈N₂O·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD30099270
CAS-Nummer: 1260646-01-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR