Ethyl (S)-4-(piperidin-2-yl)benzoate hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321267-100MG
APOSOR321267-100MGEA 168 EUR
APOSOR321267-100MG
Ethyl (S)-4-(piperidin-2-yl)benzoate hydrochloride 97%
Ethyl (S)-4-(piperidin-2-yl)benzoate hydrochloride
Formel: C₁₄H₁₉NO₂·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD30099275
CAS-Nummer: 1388117-52-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR