Ethyl (R)-3-(piperidin-2-yl)benzoate hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321270-100MG
APOSOR321270-100MGEA 168 EUR
APOSOR321270-100MG
Ethyl (R)-3-(piperidin-2-yl)benzoate hydrochloride 97%
Ethyl (R)-3-(piperidin-2-yl)benzoate hydrochloride
Formel: C₁₄H₁₉NO₂·ClH
Molecular Weight: 269,767 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099273
CAS-Nummer: 2244064-28-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR