Ethyl (S)-3-(piperidin-2-yl)benzoate hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR321271-100MG
APOSOR321271-100MGEA
333
EUR
APOSOR321271-100MG
Ethyl (S)-3-(piperidin-2-yl)benzoate hydrochloride 97%
Ethyl (S)-3-(piperidin-2-yl)benzoate hydrochloride
Formel:
C₁₄H₁₉NO₂·ClH Molecular Weight: 269,767 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30099274 CAS-Nummer: 2244064-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...