(6-Oxo-1,6-dihydropyridin-3-yl)boronic acid 97%

Lieferant: Apollo Scientific
Gefahr

OR321272-250MG
APOSOR321272-250MGEA 208 EUR
APOSOR321272-250MG
(6-Oxo-1,6-dihydropyridin-3-yl)boronic acid 97%
(6-Oxo-1,6-dihydropyridin-3-yl)boronic acid
Formel: C₅H₆BNO₃
Lagertemperatur: Kühlschrank
MDL: MFCD22123505
CAS-Nummer: 903899-13-8
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR