4-(Piperidin-3-yl)benzamide hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321273-100MG
APOSOR321273-100MGEA 168 EUR
APOSOR321273-100MG
4-(Piperidin-3-yl)benzamide hydrochloride 97%
4-(Piperidin-3-yl)benzamide hydrochloride
Formel: C₁₂H₁₆N₂O·ClH
Molecular Weight: 240,729 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099276
CAS-Nummer: 1337644-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR