5-(Piperidin-3-yl)pyridin-2-amine dihydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR321274-100MG
APOSOR321274-100MGEA
168
EUR
APOSOR321274-100MG
5-(Piperidin-3-yl)pyridin-2-amine dihydrochloride 95%
5-(Piperidin-3-yl)pyridin-2-amine dihydrochloride
Formel:
C₁₀H₁₅N₃·2ClH Molecular Weight: 250,168 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD30099277 CAS-Nummer: 2244085-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...