tert-Butyl 4-(6-chloropyridin-3-yl)-3,6-dihydro-2H-pyridine-1-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

OR321275-100MG
APOSOR321275-100MGEA 137 EUR
APOSOR321275-100MG
tert-Butyl 4-(6-chloropyridin-3-yl)-3,6-dihydro-2H-pyridine-1-carboxylate 95%
tert-Butyl 4-(6-chloropyridin-3-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
Formel: C₁₅H₁₉ClN₂O₂
Molecular Weight: 294,777 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28978570
CAS-Nummer: 1169699-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR