tert-Butyl 4-(3-cyanophenyl)-3,6-dihydropyridine-1(2H)-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

OR321276-250MG
APOSOR321276-250MGEA 137 EUR
APOSOR321276-250MG
tert-Butyl 4-(3-cyanophenyl)-3,6-dihydropyridine-1(2H)-carboxylate 95%
tert-Butyl 4-(3-cyanophenyl)-3,6-dihydropyridine-1(2H)-carboxylate
Formel: C₁₇H₂₀N₂O₂
Molecular Weight: 284,353 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD24466376
CAS-Nummer: 370864-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR