2-Methyl-4-(piperidin-3-yl)pyridine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321279-100MG
APOSOR321279-100MGEA 137 EUR
APOSOR321279-100MG
2-Methyl-4-(piperidin-3-yl)pyridine dihydrochloride 97%
2-Methyl-4-(piperidin-3-yl)pyridine dihydrochloride
Formel: C₁₁H₁₆N₂·2ClH
Molecular Weight: 249,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30099280
CAS-Nummer: 2230600-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR