2-Chloro-1-(1-methyl-1H-indol-3-yl)ethanone 97%

Lieferant: Apollo Scientific

OR32128-1G
APOSOR32128-1GEA 1570 EUR
APOSOR32128-1G
2-Chloro-1-(1-methyl-1H-indol-3-yl)ethanone 97%
2-Chloro-1-(1-methyl-1H-indol-3-yl)ethanone
Formel: C₁₁H₁₀ClNO
Molecular Weight: 207,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03848177
CAS-Nummer: 17716-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR