3-Bromo-2,4-dimethoxybenzonitrile 95%

Lieferant: Apollo Scientific
Achtung

OR321280-500MG
APOSOR321280-500MGEA 580 EUR
APOSOR321280-500MG
3-Bromo-2,4-dimethoxybenzonitrile 95%
3-Bromo-2,4-dimethoxybenzonitrile
Formel: C₉H₈BrNO₂
Molecular Weight: 242,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29040229
CAS-Nummer: 1891994-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR