(S)-4,5-Dimethyl-2-(pyrrolidin-2-yl)thiazole hydrobromide 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR321282-100MG
APOSOR321282-100MGEA
168
EUR
APOSOR321282-100MG
(S)-4,5-Dimethyl-2-(pyrrolidin-2-yl)thiazole hydrobromide 95%
(S)-4,5-Dimethyl-2-(pyrrolidin-2-yl)thiazole hydrobromide
Formel:
C₉H₁₄N₂S·BrH Lagertemperatur: Raumtemperatur |
MDL:
MFCD30828706 CAS-Nummer: 445280-79-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...