(R)-3-Methyl-5-(piperidin-2-yl)-1,2,4-oxadiazole hydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR321283-100MG
APOSOR321283-100MGEA
168
EUR
APOSOR321283-100MG
(R)-3-Methyl-5-(piperidin-2-yl)-1,2,4-oxadiazole hydrochloride 95%
(R)-3-Methyl-5-(piperidin-2-yl)-1,2,4-oxadiazole hydrochloride
Formel:
C₈H₁₃N₃O·ClH Molecular Weight: 203,669 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD30730130 CAS-Nummer: 1604285-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...