(S)-3-Methyl-5-(pyrrolidin-2-yl)isoxazole hydrobromide 95%

Lieferant: Apollo Scientific
Achtung

OR321285-100MG
APOSOR321285-100MGEA 148 EUR
APOSOR321285-100MG
(S)-3-Methyl-5-(pyrrolidin-2-yl)isoxazole hydrobromide 95%
(S)-3-Methyl-5-(pyrrolidin-2-yl)isoxazole hydrobromide
Formel: C₈H₁₂N₂O·BrH
Molecular Weight: 233,106 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30728436
CAS-Nummer: 2244064-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR