1-[6-(2,3-Dihydro-1H-isoindol-2-yl)-2H-1,3-benzodioxol-5-yl]ethan-1-one, tech

Lieferant: Apollo Scientific

OR32129-1G
APOSOR32129-1GEA 1570 EUR
APOSOR32129-1G
1-[6-(2,3-Dihydro-1H-isoindol-2-yl)-2H-1,3-benzodioxol-5-yl]ethan-1-one, tech
1-[6-(2,3-Dihydro-1H-isoindol-2-yl)-2H-1,3-benzodioxol-5-yl]ethan-1-one
Formel: C₁₇H₁₅NO₃
Molecular Weight: 281,306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787373
CAS-Nummer: 860611-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR