6-Methylpyrazolo[1,5-a]pyridine-7-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR321292-100MG
APOSOR321292-100MGEA
464
EUR
APOSOR321292-100MG
6-Methylpyrazolo[1,5-a]pyridine-7-carboxylic acid 95%
6-Methylpyrazolo[1,5-a]pyridine-7-carboxylic acid
Formel:
C₉H₈N₂O₂ Molecular Weight: 176,172 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22396405 CAS-Nummer: 1824298-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...