Methyl 6-chloro-2-oxo-2,3-dihydro-1H-benzo[d]imidazole-4-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR321293-100MG
APOSOR321293-100MGEA 168 EUR
APOSOR321293-100MG
Methyl 6-chloro-2-oxo-2,3-dihydro-1H-benzo[d]imidazole-4-carboxylate 97%
Methyl 6-chloro-2-oxo-2,3-dihydro-1H-benzo[d]imidazole-4-carboxylate
Formel: C₉H₇ClN₂O₃
Molecular Weight: 226,616 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22562211
CAS-Nummer: 1388041-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR