Ethyl 2-oxo-2-(1H-pyrazol-4-yl)acetate 97%

Lieferant: Apollo Scientific
Achtung

OR321299-100MG
APOSOR321299-100MGEA 78.3 EUR
APOSOR321299-100MG
Ethyl 2-oxo-2-(1H-pyrazol-4-yl)acetate 97%
Ethyl 2-oxo-2-(1H-pyrazol-4-yl)acetate
Formel: C₇H₈N₂O₃
Molecular Weight: 168,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16621373
CAS-Nummer: 869557-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR