N-(2,5-Dimethyl-1H-pyrrol-1-yl)furan-2-carboxamide, tech

Lieferant: Apollo Scientific

OR32130-1G
APOSOR32130-1GEA 1570 EUR
APOSOR32130-1G
N-(2,5-Dimethyl-1H-pyrrol-1-yl)furan-2-carboxamide, tech
N-(2,5-Dimethyl-1H-pyrrol-1-yl)furan-2-carboxamide
Formel: C₁₁H₁₂N₂O₂
Molecular Weight: 204,225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03305443
CAS-Nummer: 92316-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR