tert-Butyl 3-cyanopiperazine-1-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR321305-1G
APOSOR321305-1GEA 109 EUR
APOSOR321305-1G
tert-Butyl 3-cyanopiperazine-1-carboxylate 97%
tert-Butyl 3-cyanopiperazine-1-carboxylate
Formel: C₁₀H₁₇N₃O₂
Molecular Weight: 211.261 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09991479
CAS-Nummer: 859518-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR