tert-Butyl (2-aminoethyl)(ethyl)carbamate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Amino-ethyl)-ethyl-carbamic acid tert-butyl ester

OR321309-1G
APOSOR321309-1GEA 105 EUR
APOSOR321309-1G
tert-Butyl (2-aminoethyl)(ethyl)carbamate 97%
tert-Butyl (2-aminoethyl)(ethyl)carbamate
Formel: C₉H₂₀N₂O₂
Molecular Weight: 188,267 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04114276
CAS-Nummer: 105628-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR