5-((4-Methoxycyclopenta-1,3-dien-1-yl)oxy)-2-(methylthio)pyrimidine 97%

Lieferant: Apollo Scientific
Achtung

OR321312-250MG
APOSOR321312-250MGEA 250 EUR
APOSOR321312-250MG
5-((4-Methoxycyclopenta-1,3-dien-1-yl)oxy)-2-(methylthio)pyrimidine 97%
5-((4-Methoxycyclopenta-1,3-dien-1-yl)oxy)-2-(methylthio)pyrimidine
Formel: C₁₁H₁₂N₂O₂S
Molecular Weight: 236,29 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12911419
CAS-Nummer: 1245648-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR