4-(Aminomethyl)tetrahydro-2H-pyran-4-ol hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(aminomethyl)oxan-4-ol hydrochloride

OR321313-1G
APOSOR321313-1GEA 126 EUR
APOSOR321313-1G
4-(Aminomethyl)tetrahydro-2H-pyran-4-ol hydrochloride 97%
4-(Aminomethyl)tetrahydro-2H-pyran-4-ol hydrochloride
Formel: C₆H₁₄ClNO₂
Molecular Weight: 167,6339 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18072019
CAS-Nummer: 666261-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR