Ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR321318-500MG
APOSOR321318-500MGEA 230 EUR
APOSOR321318-500MG
Ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate 97%
Ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate
Formel: C₇H₁₁N₃O₂
Molecular Weight: 169,18 g/mol
Schmelzpunkt: 54…56 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03453130
CAS-Nummer: 23286-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR