2,7,7-Trimethyl-5-oxo-4-phenyl-1,4,5,6,7,8-hexahydroquinoline-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32132-1G
APOSOR32132-1GEA 1570 EUR
APOSOR32132-1G
2,7,7-Trimethyl-5-oxo-4-phenyl-1,4,5,6,7,8-hexahydroquinoline-3-carbonitrile, tech
2,7,7-Trimethyl-5-oxo-4-phenyl-1,4,5,6,7,8-hexahydroquinoline-3-carbonitrile
Formel: C₁₉H₂₀N₂O
Molecular Weight: 292,375 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00524746
CAS-Nummer: 19732-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR