Ethyl 6-amino-5-methyl-1H-indole-2-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR321321-1G
OR321321-250MG
APOSOR321321-1GEA
840
EUR
APOSOR321321-1G
APOSOR321321-250MG
Ethyl 6-amino-5-methyl-1H-indole-2-carboxylate 97%
Ethyl 6-amino-5-methyl-1H-indole-2-carboxylate
Formel:
C₁₂H₁₄N₂O₂ Molecular Weight: 218.252 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD28165861 CAS-Nummer: 1175787-04-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...