4-(2,3-Dichlorophenyl)piperidine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321334-100MG
APOSOR321334-100MGEA 546 EUR
APOSOR321334-100MG
4-(2,3-Dichlorophenyl)piperidine hydrochloride 97%
4-(2,3-Dichlorophenyl)piperidine hydrochloride
Formel: C₁₁H₁₃Cl₂N·ClH
Molecular Weight: 266.595 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD25958287
CAS-Nummer: 1354030-49-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR