2-[2-(Hydroxymethyl)phenoxy]benzenecarbonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 2-[2-(Hydroxymethyl)phenoxy]benzonitrile

OR32134-1G
APOSOR32134-1GEA 671 EUR
APOSOR32134-1G
2-[2-(Hydroxymethyl)phenoxy]benzenecarbonitrile 95%
2-[2-(Hydroxymethyl)phenoxy]benzenecarbonitrile
Formel: C₁₄H₁₁NO₂
Molecular Weight: 225,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03844844
CAS-Nummer: 59167-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR