tert-Butyl (S)-3-(4-bromophenyl)piperidine-1-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR321343-1G
OR321343-5G
APOSOR321343-1GEA
95.7
EUR
APOSOR321343-1G
APOSOR321343-5G
tert-Butyl (S)-3-(4-bromophenyl)piperidine-1-carboxylate 97%
tert-Butyl (S)-3-(4-bromophenyl)piperidine-1-carboxylate
Formel:
C₁₆H₂₂BrNO₂ Molecular Weight: 340.255 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27987933 CAS-Nummer: 1476776-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...