(1S,2S)-Boc-Acpc 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1S,2S)-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid
OR321345-5G
APOSOR321345-5GEA
917
EUR
APOSOR321345-5G
(1S,2S)-Boc-Acpc 97%
(1S,2S)-Boc-Acpc
Formel:
C₁₁H₁₉NO₄ Molecular Weight: 229,28 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04974224 CAS-Nummer: 143679-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...