4,5-Dibromo-3-methoxythiophene-2-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR321346-250MG OR321346-1G
APOSOR321346-250MGEA 502 EUR
APOSOR321346-250MG APOSOR321346-1G
4,5-Dibromo-3-methoxythiophene-2-carboxylic acid 95%
4,5-Dibromo-3-methoxythiophene-2-carboxylic acid
Formel: C₆H₄Br₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD26129618
CAS-Nummer: 120715-53-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR