2-Methyl-2-(1H-pyrazol-1-yl)propanoic acid 97%

Lieferant: Apollo Scientific

OR32135-1G
APOSOR32135-1GEA 130 EUR
APOSOR32135-1G
2-Methyl-2-(1H-pyrazol-1-yl)propanoic acid 97%
2-Methyl-2-(1H-pyrazol-1-yl)propanoic acid
Formel: C₇H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03844849
CAS-Nummer: 851975-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR