3-Carbamoylpyrrolidine-1-carboxylic acid tert-butyl ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxamide, 1-Boc-3-pyrrolidinecarboxamide, tert-Butyl 3-Carbamoylpyrrolidine-1-carboxylate, 3-Carbamoylpyrrolidine-1-carboxylic acid tert-Butyl ester, 1-Boc-3-Carbamoylpyrrolidine

OR321355-5G
APOSOR321355-5GEA 203 EUR
APOSOR321355-5G
3-Carbamoylpyrrolidine-1-carboxylic acid tert-butyl ester 97%
3-Carbamoylpyrrolidine-1-carboxylic acid tert-butyl ester
Formel: C₁₀H₁₈N₂O₃
Molecular Weight: 214.27 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04116210
CAS-Nummer: 122684-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR