4-(Boc-amino)-2-chloro-3-pyridinecarbaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl (2-chloro-3-formylpyridin-4-yl)carbamate

OR321359-1G
APOSOR321359-1GEA 212 EUR
APOSOR321359-1G
4-(Boc-amino)-2-chloro-3-pyridinecarbaldehyde 95%
4-(Boc-amino)-2-chloro-3-pyridinecarbaldehyde
Formel: C₁₁H₁₃ClN₂O₃
Molecular Weight: 256,69 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16659013
CAS-Nummer: 893423-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR