(3Z)-3-[(2-Aminophenyl)methylidene]-2,3-dihydro-1H-indol-2-one, tech

Lieferant: Apollo Scientific

OR32136-1G
APOSOR32136-1GEA 1570 EUR
APOSOR32136-1G
(3Z)-3-[(2-Aminophenyl)methylidene]-2,3-dihydro-1H-indol-2-one, tech
(3Z)-3-[(2-Aminophenyl)methylidene]-2,3-dihydro-1H-indol-2-one
Formel: C₁₅H₁₂N₂O
Molecular Weight: 236,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04124213
CAS-Nummer: 17990-71-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR