7-Amino-2,2-dimethyl-2H-benzo[b][1,4]oxazin-3(4H)-one 97%

Lieferant: Apollo Scientific
Achtung

OR321360-1G
APOSOR321360-1GEA 84.1 EUR
APOSOR321360-1G
7-Amino-2,2-dimethyl-2H-benzo[b][1,4]oxazin-3(4H)-one 97%
7-Amino-2,2-dimethyl-2H-benzo[b][1,4]oxazin-3(4H)-one
Formel: C₁₀H₁₂N₂O₂
Molecular Weight: 192,22 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11048439
CAS-Nummer: 105807-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR