Methyl 4-ethyl-2-methyl-1H-imidazole-5-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR321366-250MG
APOSOR321366-250MGEA 212 EUR
APOSOR321366-250MG
Methyl 4-ethyl-2-methyl-1H-imidazole-5-carboxylate 97%
Methyl 4-ethyl-2-methyl-1H-imidazole-5-carboxylate
Formel: C₈H₁₂N₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD12756158
CAS-Nummer: 1245644-44-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR