3-(1-Piperazinyl)-1,2-benzothiazol 97%

Lieferant: Apollo Scientific
Gefahr

OR321369-100G
APOSOR321369-100GEA 102 EUR
APOSOR321369-100G
3-(1-Piperazinyl)-1,2-benzothiazol 97%
3-(1-Piperazinyl)-1,2-benzisothiazole
Formel: C₁₁H₁₃N₃S
Lagertemperatur: Kühlschrank
MDL: MFCD04117970
CAS-Nummer: 87691-87-0
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR