2-[(2,3,4,9-Tetrahydro-1H-carbazol-1-yl)amino]ethan-1-ol, tech

Lieferant: Apollo Scientific

OR32137-1G
APOSOR32137-1GEA 1570 EUR
APOSOR32137-1G
2-[(2,3,4,9-Tetrahydro-1H-carbazol-1-yl)amino]ethan-1-ol, tech
2-[(2,3,4,9-Tetrahydro-1H-carbazol-1-yl)amino]ethan-1-ol
Formel: C₁₄H₁₈N₂O
Molecular Weight: 230,306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04124240
CAS-Nummer: 174152-39-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR